Search Thermo Fisher Scientific
The Thermo Scientific™ Finntip™ Flex™ Pipette Tips reduce the amount of force needed to attach and eject tips, significantly lowering the risk of repetitive stress injury (RSI). Ensure a better seal with the pipetter module, for higher accuracy and precision.
| Catalog Number | Volume (Metric) | Color Code | Fit for Purpose |
|---|---|---|---|
| 94060103 | |||
| 94060120 | |||
| 94060100 | |||
| 94060310 | |||
| 94060313 | |||
| 94060320 | |||
| 94060510 | |||
| 94060513 | 300 μL | Orange | Sterile & PCR Ready |
| 94060520 | |||
| 94060710 | |||
| 94060713 | |||
| 94060720 |
Finntip™ Flex™ Pipette Tips are designed to fit Finnpipette™ F1, F2 and Novus pipettes. They enable you to pipette accurately and precisely, with low liquid retention. The compatibility between the tip and the pipette is indicated with color codes. These Flex™ tips have an ergonomic design that lower the risk of RSI and use less plastic.
Key Features:
Softer Finntip Flex Tip
Proven quality
Versatile Tip Range
Immediate traceability
Note:
Availability may vary by country.