Check Order Status
Finntip™ Stepper Pipette Tips
Actual product may vary
Thermo Fisher Scientific
Finntip™ Stepper Pipette Tips
Integrate Thermo ScientificTM FinntipTM Stepper Pipette Tips with the Thermo ScientificTM FinnpipetteTM Stepper Pipetter.
Have Questions?
Change view
Catalog Number
Volume (Metric)
Sterility
9404200
5 mL
Non-sterile
9404180
1250 μL
Non-sterile
9404210
12.5 mL
Non-sterile
9404220
25 mL
Non-sterile
9404230
50 mL
Non-sterile
5 Options
Catalog number
9404200
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Volume (Metric):
5 mL
Sterility:
Non-sterile
Integrate Thermo Scientific™ Finntip™ Stepper Pipette Tips with the Thermo Scientific™ Finnpipette™ Stepper Pipetter.
Nonsterile tips are bulk packaged; sterile tips are individually wrapped
Specifications
Volume (Metric)
5 mL
Volume (Metric) Dispensing
100, 200, 300, 400, 500 μL
Filtered
Non-filtered
Sterility
Non-sterile
Certifications/Compliance
ISO 9001, ISO 14001
Format
Bulk
For Use With (Equipment)
Thermo Scientific FinnPipette Stepper
Product Line
FinnTip
Unit Size
50