Specialty Racks
Specialty Racks
Actual product may vary
Specialty Racks
Specialty Racks
Specialty Racks
Thermo Fisher Scientific
Specialty Racks
Maximize the storage space in your ultra-low temperature freezer with these racks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Holds
9200942 in. Box Racks - Sliding drawer rack13 cu. ft. capacity upright freezerspk
830032Specialty Racks - 10-compartment container17.3 and 23 cu. ft. capacity upright freezersVaries
9200173 in. Box Racks - Adjustable side access rack17.3 and 23 cu. ft. capacity upright freezersBoxes (3 in.)
9200182 in. Box Racks - Adjustable side access rack17.3 and 23 cu. ft. capacity upright freezersBoxes (2 in.)
9200222 in. Box Racks - Adjustable side access rack17.3 and 23 cu. ft. capacity upright freezersBoxes (2 in.)
9200962 in. Box Racks - Sliding drawer rack17.3 and 23 cu. ft. capacity upright freezerspk
9200973 in. Box Rack - Sliding Drawer17.3 and 23 cu. ft. capacity upright freezersBoxes (3 in.)
Catalog number 920094
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
2 in. Box Racks - Sliding drawer rack
For Use With (Equipment):
13 cu. ft. capacity upright freezers
Holds:
pk
Maximize the storage space in your ultra-low temperature freezer with these Thermo Scientific™ Racks for previous generation Forma™ 900, 7000 Series, Revco™ UxF, HERAfreeze™ HFU T and TSE/TSD Series Freezers. Includes sliding drawer racks, adjustable side access box racks and microplate racks in standard or deepwell formats.
  • With sliding drawer racks
  • Adjustable side access box racks
  • Microplate racks in standard or deepwell formats
  • Warranty varies by country, please contact us for details
Specifications
Product Type2 in. Box Racks - Sliding Drawer
Description2 in. Box Racks - Sliding drawer rack
Holdspk
ShapeSquare
For Use With (Equipment)13 cu. ft. capacity upright freezers
Width (Metric)14 cm
Width (English)5.5 in.
Height (Metric)42.2 cm
Height (English)16.6 in.
Depth (English)11.9 in.
Depth (Metric)30.2 cm
Unit Size