Accessories for Digital Vortex Mixers
Accessories for Digital Vortex Mixers
Actual product may vary
Thermo Fisher Scientific
Accessories for Digital Vortex Mixers
Get assorted accessories for Digital Vortex Mixers
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionDimensions (L x W)Material
8888212319 x 10 mm dia.Tube Holder87.5 x 40 mmFoam, plastic
88882122Microplate Tray136.1 x 97.3 mmPlastic
8888212413 x 12 mm dia. Tube Holder87.5 x 40 mmFoam, plastic
88882126
also known as 88882126 
7 x 20 mm dia. Tube Holder87.5 x 55 mmFoam, plastic
888821274 x 26 mm dia.Tube Holder87.5 x 55 mmFoam, plastic
Catalog number 88882123
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
19 x 10 mm dia.Tube Holder
Dimensions (L x W):
87.5 x 40 mm
Material:
Foam, plastic
Assorted accessories for Digital Vortex Mixers
Specifications
Tube Size10 mm dia.
Product TypeTube Holder
MaterialFoam, plastic
Dimensions (L x W)87.5 x 40 mm
ColorBlack
Description19 x 10 mm dia.Tube Holder
For Use With (Equipment)Digital Vortex mixer
Unit Size