Check Order Status
Accessories for Digital Mini Rotators
Actual product may vary
Thermo Fisher Scientific
Accessories for Digital Mini Rotators
Get assorted accessories for use with Digital Mini Rotators
Have Questions?
Change view
Catalog Number
Description
Dimensions (L x W)
88882110
1 L Flask Clamp
130 x 77 mm
88882105
Universal (Aluminum) Platform
310 x 284 mm
88882104
Universal Attachment
307 x 303 mm
3 Options
Catalog number
88882110
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Description:
1 L Flask Clamp
Dimensions (L x W):
130 x 77 mm
Assorted accessories for use with Digital Mini Rotators
Specifications
Product Type
1 L Flask Clamp
Material
Stainless Steel, rubber
Dimensions (L x W)
130 x 77 mm
Description
1 L Flask Clamp
For Use With (Equipment)
Digital Mini Rotators, Rocker, Rotator, Mixer
Unit Size