Check Order Status
Accessories for Digital Mini Rotators
Actual product may vary
Thermo Fisher Scientific
Accessories for Digital Mini Rotators
Get assorted accessories for use with Digital Mini Rotators
Have Questions?
Change view
Catalog Number
Description
Dimensions (L x W)
88882104
Universal Attachment
307 x 303 mm
88882105
Universal (Aluminum) Platform
310 x 284 mm
88882110
1 L Flask Clamp
130 x 77 mm
3 Options
Catalog number
88882104
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Description:
Universal Attachment
Dimensions (L x W):
307 x 303 mm
Assorted accessories for use with Digital Mini Rotators
Specifications
Material
Stainless Steel, plastic, rubber
Dimensions (L x W)
307 x 303 mm
Dimensions (L x W x H)
307 x 303 x 145 mm
Description
Universal Attachment
For Use With (Equipment)
Digital Mini Rotators, Rocker, Rotator, Mixer
Unit Size