Racks and Accessories for CO2 Resistant Shaker
Racks and Accessories for CO<sub>2</sub> Resistant Shaker
Actual product may vary
Thermo Fisher Scientific
Racks and Accessories for CO2 Resistant Shaker
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
Have Questions?
Catalog NumberIncludes
88881122
Catalog number 88881122
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Mix and match different size accessories on a single platform with flexible accessories for the Thermo Scientific™ CO2 resistant shaker.
  • Compatible with Thermo Scientific™ CO2 Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
  • Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.
Specifications
Product TypeUniversal Platform
MaterialAluminum
Dimensions (L x W)360 x 300 mm
For Use With (Equipment)CO2 Resistant Shaker
Unit Size