Block, 28 x 10 mm dia., EMD
Product Image
Thermo Fisher Scientific
Block, 28 x 10 mm dia., EMD
For Use With: Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Have Questions?
Catalog number 88870102
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Specifications
Description28 x 10mm dia. block
For Use With (Equipment)Thermo Scientific™ Digital and Touch Screen Dry Baths/Block Heaters (No compact models)
Holds28 x 10mm
TypeBlock
Dimensions (L x W x H)76 x 124 x 39 mm (3 x 4.9 x 1.5 in.)
Unit Size