Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Actual product may vary
Thermo Fisher Scientific
Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Speed up your analysis with high-throughput measurements.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeHolds
840-3036004-position Carousel Base4 x 10mm Pathlength Cells
840-314700Rect Cell 4-position Carousel4 x 50mm PL Rectangular Cells
840-3034008-position Carousel8 x 10mm Pathlength Cells
Catalog number 840-303600
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Type:
4-position Carousel Base
Holds:
4 x 10mm Pathlength Cells
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.
Specifications
Type4-position Carousel Base
For Use With (Equipment)GENESYS 140/150/180, BioMate 160 Spectrophotometers
Holds4 x 10mm Pathlength Cells
Unit Size