Check Order Status
Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Actual product may vary
Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers
Organize vial storage within cryo boxes with these fiberboard box dividers.
Have Questions?
Change view
Catalog Number
Holds
Quantity
820016
16 (4 x 4 Grid)
Each
FIS13-994-085
25 (5 x 5 Grid)
Each
2 Options
Catalog number
820016
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Holds:
16 (4 x 4 Grid)
Quantity:
Each
Organize vial storage with Thermo Scientific™ Fiberboard Box Dividers for Ultra-Low Temperature and Cryogenic Freezers.
Use with fiberboard storage boxes to divide box into grids for vial storage
Box dividers for cold storage application available in 7 x 7 , 8 x 8, 9 x 9 or 10 x 10 grids
Specifications
Quantity
Each
Holds
16 (4 x 4 Grid)
Unit Size