Windshielded Rotor for Cryofuge™ 6000i/8500i
Windshielded Rotor for Cryofuge™ 6000i/8500i
Actual product may vary
Thermo Fisher Scientific
Windshielded Rotor for Cryofuge™ 6000i/8500i
Increase lab productivity with the Windshielded Rotor for Cryofuge 6000i/8500i, which comes with a lid and without buckets.
Have Questions?
Catalog NumberCapacity
750066066 x 1000 mL
Catalog number 75006606
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity:
6 x 1000 mL
Increase lab productivity with the Thermo Scientific™ Windshielded Rotor for Cryofuge™ 6000i/8500i, which comes with a lid and without buckets.
Specifications
DescriptionWindshielded Swinging Bucket Rotor
MaterialStainless Steel Rotor Body, Aluminum Bucket
TypeSwinging Bucket Rotor
Capacity6 x 1000 mL
Max. RCF6,010 x g (Round), 5,312 x g (Double Blood Bag), 4,984 x g (Double Rectangular)
Max. Speed4,020 rpm (5,050 rpm with Cryofuge 6000i, Cryofuge 8500i)
K Factor10,279
For Use With (Equipment)Thermo Scientific Cryofuge 6000i and 8500i Large Capacity Centrifuges
Angle90°
Unit Size