Shelves for Heratherm™ Ovens and Incubators
Shelves for Heratherm™ Ovens and Incubators
Actual product may vary
Thermo Fisher Scientific
Shelves for Heratherm™ Ovens and Incubators
Perforated and wire mesh shelves
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDepth (English)Width (English)For Use With (Equipment)Length (English)
5012777719.8 in.19.8 in.Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l17.2 in.
5012777114.8 in.14.8 in.Heratherm General Protocol Microbiological Incubator 100L12.9 in.
5012777212.9 in.12.9 in.Heratherm General Protocol Microbiological Incubator 180l12.9 in.
5012777014.8 in.14.8 in.Heratherm General Protocol Microbiological Incubator 60l12.9 in.
5012777412.9 in.
5012776319.8 in.19.8 in.Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l17.2 in.
5013524121.06 in.
5013524221.06 in.
5013524521.06 in.21.06 in.Heratherm Advanced Protocol Oven 400I OMH 400 and General Protocol Incubator 400l IGS400
5013524621.06 in.
Catalog number 50127777
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Depth (English):
19.8 in.
Width (English):
19.8 in.
For Use With (Equipment):
Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l
Length (English):
17.2 in.
Perforated and wire mesh shelves
Specifications
For Use With (Equipment)Heratherm Advanced Protocol/Advanced Protocol Security Microbiological Incubator 180l
TypeShelf
Includes1 Shelf, 2 shelf supports
MaterialStainless Steel
No. of Shelves1
Length (English)17.2 in.
Width (English)19.8 in.
Width (Metric)50.2 cm
Depth (English)19.8 in.
Depth (Metric)50.2 cm
Unit Size