Calibrate airborne optical particle counters to comply with ISO 21501-4 with Thermo Scientific™ 4D Series Dry Microsphere Size Standards.
Have Questions?
Change view
Catalog Number
Diameter (Metric)
4D-03
3 μm
4D-10
10 μm
2 Options
Catalog number 4D-03
View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
3 μm
Calibrate airborne optical particle counters (OPC) to comply with ISO 21501-4 regulations with the Thermo Scientific™ 4D Series Dry Microsphere Size Standards. Ideal for instrument manufacturers, as well as QA/QC managers in pharmaceutical, biotech and electronics manufacturing, these microsphere reference beads feature a calibration uncertainty—the range of values within which the actual mean diameter will lie—that is less than or equal to 2.5% of the mean diameter.
The diameter is traceable to the National Institute of Standards and Technology (NIST), and the product’s Certificate of Calibration and Traceability provides the documentation needed for QC/regulatory programs and audits.
Mean size standard uncertainty of ≤2.5% enables calibration of air particle counters to ISO
Superior uniformity provides exact calibration point when setting the calibration curve
Diameter is traceable to NIST, a known government standard
Certificate of Traceability provides objective evidence for QC compliance
Available in 3μm, 5μm, 7μm and 10μm diameters
Conveniently packaged in dropper-tipped bottles for direct dispensing into sampling chamber
Specifications
AdditiveTrace flow agent
Certifications/ComplianceCertificate of Calibration and Traceability to NIST, MSDS
Concentration100% solids
Contents1g
ColorWhite
For Use With (Application)Airborne Optical Particle Counter Calibration
MaterialPolystyrene Divinylbenzene (PS-DVB)
Product TypeSize Standard Particles
Refractive Index1.59 at 589nm (25°C)
Diameter (Metric)3 μm
Unit Size
Contents & Storage
Unless otherwise stated, store at ambient room temperature. Store upright; keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.