250mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 12
250mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 12
Actual product may vary
250mL Erlenmeyer (Shaker) Flask, Single-Use PETG, Plain Bottom, Filter Cap, Case of 12
Save preparation time and avoid contamination risk with these single-use flasks, ideal for suspension of cell culture, media preparation, mixing and storage.
Have Questions?
Catalog number 4115-0250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Save preparation time and avoid contamination risk with Thermo Scientific™ Nalgene™ Single-Use PETG Erlenmeyer Flasks with Plain Bottom, ideal for suspension of cell culture, media preparation, mixing and storage. Crystal clear PETG is robust and break resistant, increasing personal safety. All sizes are available vented or non-vented.
Features:
  • Sterile and single use: Reduce contamination risk
  • Collapses when autoclaved: Reduce bio hazardous waste
  • Graduated for quick volume assessment
Specifications
Capacity (English)8 oz.
Capacity (Metric)250 mL
MaterialPETG
No. per Case12
No. per Pack1
ShapeErlenmeyer
Surface CoatingNone
AutoclavableNot Autoclavable
DisposableSingle-use
GraduatedYes
Mouth StyleNarrow
SterilitySterile
Bottom ShapePlain
Closure TypeVented
Closure Size38-430 mm
Unit Size