Check Order Status
Forma™ Freezer Replacement Shelves
Actual product may vary
Forma™ Freezer Replacement Shelves
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
Have Questions?
Change view
Catalog Number
For Use With (Equipment)
Type
224134
13.0 cu. ft. Freezers
Shelf Kit
224135
17.3 cu. ft. Freezers
Shelf Kit
224158
23.0 cu. ft. Freezers
Shelf Kit
3 Options
Catalog number
224134
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
13.0 cu. ft. Freezers
Type:
Shelf Kit
Use Thermo Scientific™ Forma™ Freezer Replacement Shelves with Forma 8600 and 900 Series Upright Freezers.
Stainless steel
Kits include mounting hardware
Specifications
Type
Shelf Kit
Capacity (English)
13 cu. ft.
Capacity (Metric)
368.1 L
Style
Upright
Color
Silver
For Use With (Equipment)
13.0 cu. ft. Freezers
Unit Size