Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Actual product may vary
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure
Conserve space with these square, narrow-mouth HDPE bottles that are chemically and break resistant, as well as economically priced for a wide range of lab uses.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)dimClosureSize
2018-10001 L
2018-006060 mL
2018-0500500 mL
FKS10637711500 mL
Catalog number 2018-1000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Capacity (Metric):
1 L
Conserve space with Thermo Scientific™ Nalgene™ Square Narrow-Mouth HDPE Bottles with Closure. HDPE bottles are the most chemically and break resistant of all Nalgene square bottles, yet are economically priced for a wide range of lab uses. They are made only from high-quality, laboratory-grade plastic materials for dependably low leachables and extractables. This product is ACT Labeled by My Green Lab to enable data-based sustainable product purchasing decisions.
Features:
  • Square sides allow for good use of space and an easy grip
  • Caps provide leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
  • Graduated for easy dispensing
  • Leakproof† when used with Nalgene closures
  • Can be used to -100°C (-148°F)
  • Not autoclavable
  • Participates in My Green Lab's ACT Label program for Accountability, Consistency and Transparency (ACT) around evaluating product sustainability
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Includes:
Leakproof† Polypropylene Screw Caps
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually.  Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
† The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Specifications
Closure MaterialPolypropylene
Closure Size38-430 mm
Closure TypeScrew Cap
Diameter (Metric)92 mm
Height (English)8.6 in.
Height (Metric)218 mm
Through the Closure DispensingNo
Width (English)3.9 in.
Product TypeNarrow Mouth Bottle
Capacity32 oz., 1000 mL
Capacity (Metric)1 L
Capacity (English)32 oz.
ColorTranslucent
DescriptionNarrow-Mouth Square Bottle
Diameter (English)3.7 in.
Diameter (English) Inner Neck1.1 in.
Diameter (Metric) Inner Neck28 mm
IncludesLeakproof polypropylene screw closures
MaterialHDPE
No. per Case24
No. per Pack6
SterilityNon-sterile
Closure ColorNatural, translucent
GraduationsYes
Certifications/ComplianceUSP Class VI; FDA - 21 CFR
Closure IncludedYes
AutoclavableNot Autoclavable
GraduatedYes
MouthNarrow
ShapeSquare
ACT LabelYes
Unit Size24