QSP Filter and Non-Filtered Pipette Tips
QSP Filter and Non-Filtered Pipette Tips
Actual product may vary
Thermo Fisher Scientific
QSP Filter and Non-Filtered Pipette Tips
Filter and non-filter pipette tips in a complete portfolio for your evolving liquid transfer needs.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionVolume (Metric)Tip StyleSterility
104-QQSP 10, Non-Filtered10 μLMicroPointNon-sterile
095-QQSP 5000, Non-Filtered5000 μLMacroNon-sterile
112NXL-QQSP 1000, Non-Filtered1250 μLExtended LengthNon-sterile
T090-QQSP 200, Non-Filtered200 μLBeveledNon-sterile
112XL-QQSP 1000, Non-Filtered1000 μLExtended LengthNon-sterile
TF140-200-QQSP 200, Filtered200 μLMicroPointSterile
TF112-1000-QQSP 1000, Filtered100 to 1000 μLExtended LengthSterile
TF113-20-QQSP 20, Filtered20 μLBeveledSterile
TF104-10-QQSP 10, Filtered10 μLMicroPointSterile
TF113-100-QQSP 100, Filtered100 μLBeveledSterile
Catalog number 104-Q
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
QSP 10, Non-Filtered
Volume (Metric):
10 μL
Tip Style:
MicroPoint
Sterility:
Non-sterile
Thermo Scientific™ QSP filter and non-filtered pipette tips are made from 100% virgin polypropylene utilizing state of the art molds and molding methods. Pipette tips are molded with high clarity resin and many include graduation marks and micro point design. Packaging options range from bulk to rack and rack sterile and include environmentally friendly options like tip reload systems and stack racks.
Featuring:
  • Micro point design tip orifice
  • High clarity resin
  • Graduation marks
  • Universal fit
  • Small micron filter
  • Modern and robust tip rack design
Clean Claims:
QSP Pipette Tips are free of:
  • Non-Sterile:
    • RNase/DNase
    • DNA
    • Pyrogen
  • E-Beam Sterilized:
    • RNase/DNase
    • DNA
    • Pyrogen/Endotoxin
    • Bioburden
    • PCR inhibitors
    • Endotoxin ATP
To order a clean claim lot-specific certification, contact your local dealer with the Cat. No and Lot No.
Recommend using with Finnpipette Digital pipette, but also compatible with other brands of pipettes.
Specifications
DescriptionQSP 10, Non-Filtered
Volume (Metric)10 μL
FilteredNon-filtered
SterilityNon-sterile
ColorNatural
Length (Metric)3.1cm
Clean ClaimsDNase, RNase, DNA, and Pyrogen Free
FormatBagged
Tip StyleMicroPoint
For Use With (Equipment)Universal Fit
Product LineQSP
Unit Size1000 TIPS X 1BAG