Pretreatment Cartridges and Filters
Pretreatment Cartridges and Filters
Actual product may vary
Pretreatment Cartridges and Filters
Pretreatment Cartridges and Filters
Thermo Fisher Scientific
Pretreatment Cartridges and Filters
Choose one of these Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your point of use water purification system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
06.545210 ft. (25.4 cm) Hardness stabilizing cartridge. For use with 09.4000
09.4000Double cartridge pretreatment system (includes 5µm filter with carbon cartridge and a hardness stabilizer cartridge)
06.52045µm filter with hardness stabilizer
09.4003Pretreatment filter for the Smart2Pure system. Includes filter housing and 1µm filter
501340225µm filter with carbon with 10 ft. filter housing
Catalog number 06.5452
Description:
10 ft. (25.4 cm) Hardness stabilizing cartridge. For use with 09.4000
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
Description10 ft. (25.4 cm) Hardness stabilizing cartridge. For use with 09.4000
For Use With (Equipment)LabTower TII, Pacific RO, Pacific TII
TypeStabilizing Cartridge
Height (English)10 ft.
Height (Metric)25.4 cm
Unit Size