Sorvall™ High Performance Centrifuge Bottle
Sorvall™ High Performance Centrifuge Bottle
Actual product may vary
Sorvall™ High Performance Centrifuge Bottle
Sorvall™ High Performance Centrifuge Bottle
Thermo Fisher Scientific
Sorvall™ High Performance Centrifuge Bottle
Designed for superspeed and large capacity centrifuge performance up to 20,584 x g, this high-performance one liter centrifuge bottle boosts productivity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialClosure Included
010-1455PPCONo
010-1456PPCOYes
Catalog number 010-1455
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Material:
PPCO
Closure Included:
No
Designed for superspeed and large capacity centrifuge performance up to 20,584 x g, the Thermo Scientific™ Sorvall™ high-performance 1 liter centrifuge bottle boosts productivity, and the new high-strength aluminum cap enhances bottle support and improves chemical resistance to Phenol solutions.
Designed for use with Thermo Scientific™ Sorvall™ superspeed and large capacity centrifuges with Thermo Scientific™ Fiberlite™ 1-liter rotors, Sorvall high performance 1 liter centrifuge bottles provide:
  • One full liter in each bottle at g-forces up to 20,584 x g
  • When compared with Noryl™ plastic caps, the high-strength aluminum cap
    • Enhances bottle support
    • Improves chemical resistance to Phenol solutions
These bottles are available in both polypropylene copolymer (PPCO) bottles designed for excellent chemical resistance and are autoclavable or polycarbonate (PC) bottles designed for excellent clarity.
Based on internal testing data.
Maximum RCF
  • 17,568 x g (with Thermo Scientific Fiberlite F9-6x1000 LEX rotor in the Thermo Scientific Sorvall LYNX 6000 superspeed centrifuge)
  • 20,584 x g (Thermo Scientific Fiberlite F10-4x1000 LEX rotor in the Sorvall LYNX 6000 or 4000 superspeed centrifuge)
  • 9,333 x g (Thermo Scientific Fiberlite F5-10x1000 LEX rotor in the Thermo Scientific Sorvall RC BIOS 10 centrifuge)
  • PPCO and PC bottles: Replace bottles and plugs by 50 cycles; Replace o-rings more frequently if needed
Autoclaving
  • Aluminum cap: 121°C for 20 minutes or 134°C for 3 minutes
  • NOTE: Do not autoclave bottles with closures engaged to prevent collapse of bottles when cooling.
  • NOTE: If autoclaved, PC bottles are weakened and their useful life is shortened.
1000mL superspeed bottles are a wearable part with a finite performance life. Bottles used with compatible samples would be expected to show physical signs of wear with repeated use. Remove bottles from use which show signs of wear, cracks, crazing, discoloration, yellowing, deformation, surface scratches, abrasions or chemical attack.
Cycle Life
  • Aluminum cap: 1000 cycles or 5 years (whichever comes first)
  • PPCO and PC bottles: Replace bottles and plugs by 50 cycles; Replace o-rings more frequently if needed
Autoclaving
  • Aluminum cap: 121°C for 20 minutes or 134°C for 3 minutes
  • NOTE: Do not autoclave bottles with closures engaged to prevent collapse of bottles when cooling.
  • NOTE: If autoclaved, PC bottles are weakened and their useful life is shortened.
1000mL superspeed bottles are a wearable part with a finite performance life. Bottles used with compatible samples would be expected to show physical signs of wear with repeated use. Remove bottles from use which show signs of wear, cracks, crazing, discoloration, yellowing, deformation, surface scratches, abrasions or chemical attack.
Specifications
Capacity (Metric)1000 mL
Capacity (English)33.81 oz.
ColorClear
Fill Volume Range800 to 1000mL
MaterialPPCO
Max. Sample Density 1.2g/mL
No. per Pack2
Product LineSorvall
Closure IncludedNo
AutoclavableAutoclavable
GraduatedNo
Max. Force20,584 x g (with Thermo Scientific Fiberlite F10-4x1000 LEX rotor in the Sorvall LYNX 6000 or 4000 superspeed centrifuge)
ShapeRound
Product TypeCentrifuge Bottle
For Use With (Equipment)Super Speed and Large Capacity Centrifuges
Closure MaterialPolypropylene
Closure TypeSealing Cap
Through the Closure DispensingYes
Unit Size