Coalesces exhaust oil mist from rotary vane vacuum pump and drains back into oil case to reduce oil consumption and enable a clean work space. Exhaust can be directed into existing fume hood. Filters to 0.3 μm particles.
Have Questions?
Catalog Number
Type
WEL1417E
Exhaust Filter (Connection: NW16)
Catalog number WEL1417E
Price (MYR)
1,821.00
EA
Add to cart
Type:
Exhaust Filter (Connection: NW16)
Price (MYR)
1,821.00
EA
Add to cart
Exhaust can be directed into existing fume hood
Reduces pump noise
Use 1417E for pumps with NW16 Exhaust Connections (such as Welch CRVPro2, 4, 6, 8).
Use 1417F for pumps with NW25 Exhaust Connections (such as Welch CRVpro16, 24, 30).
Use 1417H-01 for replacement element for 1417F
Specifications
For Use WithPumps with NW16 Exhaust Connections (Welch CRVPro2, 4, 6, 8)