Laboratory Vacuum System 105 T -10ef+
Product Image
Laboratory Vacuum System 105 T -10ef+
Laboratory Vacuum System 105 T -10ef+ - Faster, better quality distillation and solvent concentration.
Have Questions?
Catalog number WEL124184
Price (MYR)
44,102.00
EA
Add to cart
Price (MYR)
44,102.00
EA
Add to cart
  • Faster, better quality distillation and solvent concentration
  • The LVS 105 T -10 ef+ system is specially designed for solvent distillation/evaporation applications and comprises of a chemical duty diaphragm pump and VCpro 600 series digital controller with rpm motor speed control (ecoflex technology).
  • Liquid separator and exhaust emissions condenser for optimal solvent recovery are also included.
  • Designed for chemical applications all wetted parts are made of high quality chemically resistant materials and clear plastic coated glassware.
  • Ideal for evaporation of high and low boiling point solvents.
  • The chemical resistant diaphragm pump (MPC series) provides a deep stable vacuum.
Specifications
Dimensions (L x W x H)250 x 260 x 435 mm
MaterialPTFE, PEEK
ModelLVS 105 T -10 ef+
Weight (Metric)9.5 kg
Flow Rate18 L/min.
For Use WithEvaporation of High and Low Boiling Point Solvents
Frequency50/60 Hz
Pump TypeVacuum System
Maximum Pressure2 mbar
Voltage115/230 V
Unit SizeEA