Search Thermo Fisher Scientific
| Catalog Number | Drive Size |
|---|---|
| WEL1012 | 10 x 12 mm |
| WEL1002 | 7 x 9 mm |
| WEL1018 | 10 x 14 mm |
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.