Double Box Wrench (Metric)
Double Box Wrench (Metric)
Actual product may vary
Double Box Wrench (Metric)
AMPCO™ Double Box Wrench performs in high-risk and demanding environments.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDrive Size
WEL10027 x 9 mm
WEL101210 x 12 mm
WEL101810 x 14 mm
Catalog number WEL1002
Price (MYR)
105.20
EA
Add to cart
Drive Size:
7 x 9 mm
Price (MYR)
105.20
EA
Add to cart
  • Made of AMPCO invented, Factory Mutual approved alloy
  • Designed for use near flammable liquids, vapors, explosive dust, sensitive electronics, magnetic equipment, batteries and in laboratories.
  • Non-Sparking
  • Non-Magnetic
  • Corrosion Resistant
Common applications include maintenance repair operations (MRO) around flammable liquids, dusts and vaports. Non-magnetic environments for MRI maintenanance and other precision magnetic and radar equipment; Laboratories, food and beverage processing, pharmaceuticals and chemicals handling and processing where corrosion resistant hand tools are needed.
Specifications
Product TypeDouble Box Wrench
Drive Size7 x 9 mm
MaterialAluminum Bronze
Certifications/ComplianceFactory Mutual (FM) Approved
Unit SizeEA