Male-Threaded Nuts
Male-Threaded Nuts
Actual product may vary
Male-Threaded Nuts
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)Length (English)MaterialWidth (English)
U-400X1/16 in. O.D. Tubing0.4 in.Stainless Steel0.25 in.
HYPU-400X1/16 in. O.D. Tubing0.4 in.Stainless Steel0.25 in.
Catalog number U-400X
View Price:Sign in to see your account pricing. Need an account? Register with us today.
For Use With (Equipment):
1/16 in. O.D. Tubing
Length (English):
0.4 in.
Material:
Stainless Steel
Width (English):
0.25 in.
Combine with Super Flangeless™ ferrule for a high pressure holding flat-bottom fitting assembly
  • Eliminates tubing twist that can loosen connections
  • Resistant to loosening through vibration
Specifications
Diameter (English) Outside0.0625 in.
Length (English)0.4 in.
Width (English)0.25 in.
For Use With (Equipment)1/16 in. O.D. Tubing
MaterialStainless Steel
TypeNut
Unit Size