Check Order Status
DropSafe ErgoLance Micro Flow Lancet
Actual product may vary
DropSafe ErgoLance Micro Flow Lancet
Sterile, single-use devices designed for the sampling of capillary blood from the fingertip of patients.
Have Questions?
Change view
Catalog Number
Color
Needle Gauge
TYL8481
Green
30G
TYL8482
Blue
25G
2 Options
Catalog number
TYL8481
Price (MYR)
313.00
Add to cart
Color:
Green
Needle Gauge:
30G
Price (MYR)
313.00
Add to cart
Special lancet shape distracts attention from needle stick
Concave housing for comfortable finger recess maximizes economy
No release button
Gentle pressure activation
Ribbed protective cap guarantees needle sterility and gives a secure grip for easy twist-off removal
Specifications
Color
Green
For Use With (Application)
Lancet for blood testing
Length (Metric)
1.5 mm
Material
Medical sharp
Needle Gauge
30G
Quantity
100 ct. per box
Product Type
Safety Lancet
Unit Size