Check Order Status
Glass pH Electrode
Actual product may vary
Glass pH Electrode
Have Questions?
Change view
Catalog Number
pH Range
Reference Electrolyte
Temperature (Metric) Operating
TYL5207
0 to 14
CRYSOLYT A
0°C to 80°C
TYL5208
0 to 14
CRISOLYT
0°C to 80°C
2 Options
Catalog number
TYL5207
Price (MYR)
956.00
Add to cart
pH Range:
0 to 14
Reference Electrolyte:
CRYSOLYT A
Temperature (Metric) Operating:
0°C to 80°C
Price (MYR)
956.00
Add to cart
Specifications
Material
Glass
pH Range
0 to 14
Temperature (Metric) Operating
0°C to 80°C
Type
pH Electrode
For Use With (Application)
Paper, fabric, leather, agar, leaves, etc
Reference Electrolyte
CRYSOLYT A
Temperature (Metric) Range
0°C to 80°C
Unit Size