Search Thermo Fisher Scientific
Catalog Number | Pore Size |
---|---|
TYL5194 | 2 mm |
FIS04-881-10G | 2 mm |
TYL5197 | 1.18 mm |
FIS04-881-10L | 1 mm |
TYL5206 | 250 μm |
FIS04-881-10Z | 106 μm |
FIS04-881-10BB | 75 μm |
TYL5213 | 75 μm |
Stainless Steel. Aperture sizes from 5.00 in. opening to 500 mesh.
All sieves are serialized and come with a Certificate of Compliance.
If Test Certificate is required (Fisher Cat. No. 04-909-16), it must be orderd along with the sieve.
The list below does not represent the complete ASTM E 11 specifications; other sizes are available. Contact your Fisher representative for more information.