Sentino™ Filters Funnel, Gamma Irradiated
Sentino™ Filters Funnel, Gamma Irradiated
Actual product may vary
Sentino™ Filters Funnel, Gamma Irradiated
Sentino™ Filter Funnels are single-use funnels designed for use with the Sentino™ pump with a press-fit seal, membrane guides and forceps access point allowing for simple set up and membrane removal.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Quantity
TYL4870100 mL25/Sleeve, 100/Pk.
CRG4870100 mL25/Sleeve, 100/Pk.
CRG4871250 mL20/Sleeve, 80/Pk.
TYL4871250 mL20/Sleeve, 80/Pk.
Catalog number TYL4870
Price (MYR)
849.00
Add to cart
Capacity (Metric):
100 mL
Quantity:
25/Sleeve, 100/Pk.
Price (MYR)
849.00
Add to cart
  • For research use only
Specifications
Capacity (Metric)100 mL
MaterialPolypropylene
Quantity25/Sleeve, 100/Pk.
SterilitySterile
Product TypeFiltering Funnel
Unit Size