Acrodisc™ Syringe Filters with PVDF Membrane and Minispike Outlet
Acrodisc™ Syringe Filters with PVDF Membrane and Minispike Outlet
Actual product may vary
Acrodisc™ Syringe Filters with PVDF Membrane and Minispike Outlet
Hydrophilic membrane compatible with a wide variety of solvents.
Have Questions?
Catalog NumberPore SizeQuantity
TYL45450.45 μm1000/Pk.
Catalog number TYL4545
Price (MYR)
849.00
Add to cart
Pore Size:
0.45 μm
Quantity:
1000/Pk.
Price (MYR)
849.00
Add to cart
  • Useful for a wide range of applications, including aggressive and non-aggressive solvent-based mobile phase
Specifications
Material (Membrane)PVDF
Pore Size0.45 μm
SterilityNon-sterile
Filtration Area1 cm2
Hold up Volume (After Air Purge)<14 μL
Housing MaterialPolypropylene
Outlet StyleMinispike Outlet
Quantity1000/Pk.
Temperature (Metric) Max55°C
Diameter (Metric)13 mm
Unit Size