Stainless Steel Basket Wire For Autoclaves
Stainless Steel Basket Wire For Autoclaves
Actual product may vary
Stainless Steel Basket Wire For Autoclaves
Suitable for waste.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
TTLBSK507-00015050 Vertical Autoclave
TTLBSK411-00052840 Bench Autoclave
TTLBSK385-00023850 Bench Autoclave
TTLBSK507-00105075 Bench Autoclave
Catalog number TTLBSK507-0001
Price (MYR)
3,170.00
Add to cart
For Use With (Equipment):
5050 Vertical Autoclave
Price (MYR)
3,170.00
Add to cart
For over 90 years Tuttnauer, as a family owned business, has been an industry leader satisfying customer expectations with top quality, high performance products and a dedicated service support team. Tuttnauer's sterilization & infection control products are trusted at over 350,000 installations worldwide including Laboratories, Pharmaceutical Facilities, Hospitals and Clinics.
  • Stainless Steel Basket Wire for bench or vertical autoclave
  • Suitable for waste
Specifications
For Use With (Application)Liquid Sterilization, Pipette and Glass Sterilization, Instrument Sterilization (Wrapped or Unwrapped), Biohazard and Waste Sterilisation, Agar Preparation, Specialized Customized Cycles.
For Use With (Equipment)5050 Vertical Autoclave
Height (Metric) Exterior250 mm
Length (Metric)500 mm
ModelVertical
Unit Size