Reacti-Therm™ Thermometers
Reacti-Therm™ Thermometers
Actual product may vary
Reacti-Therm™ Thermometers
Use these thermometers to measure temperature in dry block heaters of Thermo Scientific™ Reacti-Therm™ Heating and Stirring module.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberOperating Temperature (Metric)
TPRTS-189140°C to 100°C
TPRTS-189150°C to 200°C
Catalog number TPRTS-18914
Price (MYR)
1,042.00
EA
Add to cart
Operating Temperature (Metric):
0°C to 100°C
Price (MYR)
1,042.00
EA
Add to cart
Measure temperature in dry block heaters of the Thermo Scientific™ Reacti-Therm™ Heating and Stirring module and other laboratory equipment using Thermo Scientific™ Reacti-Therm™ Thermometers. The thermometers are mercury-free and alcohol-filled for greater safety. PTFE coating ensures that glass is impervious to corrosive materials. They are available in two different temperature ranges.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
TypeThermometer
Operating Temperature (Metric)0°C to 100°C
For Use With (Equipment)Dry Block Heaters and Other Instruments
Unit SizeEA