Reacti-Vial Reaction Vials
Product Image
Reacti-Vial Reaction Vials
An internal cone makes small sample handling easy and convenient
Have Questions?
Catalog NumberColor
TPRTS-13223Clear
Catalog number TPRTS-13223
Price (MYR)
2,578.00
PK
Add to cart
Color:
Clear
Price (MYR)
2,578.00
PK
Add to cart
  • Extra thick glass wall magnifies the sample, making these vials ideal for observing chemical reactions
  • Amber vials available for light-sensitive compounds
  • Supplied complete with Open-Top screw caps and PTFE/Rubber discs
Residue isolation, Derivative preparation, Maximum sample retrieval, Moisture protection, Sample storage, Precipitations, Centrifugations, Solvent evaporation
Specifications
ColorClear
MaterialSilica Glass
Volume (Metric)5 mL
Thread StyleInternal
Volume (Metric) Total5 mL
Closure TypeScrew Cap
Certifications/ComplianceCertificate of Analysis (COA)
TypeVial
Unit SizePK