Specifications
ChlorinatedYes
Cleanroom ClassClass 100 (ISO 5)
Accelerator FreeYes
AQL1.5
ColorWhite
DisposableSingle-use
Exterior FinishPartially Textured
Finger Thickness (English)5 mil
Finger Thickness (Metric)0.15 mm
HandAmbidextrous
Latex FreeYes
MaterialNitrile
Palm Thickness (English)4 mil
Palm Thickness (Metric)0.12 mm
PowderedPowder Free
Product TypeControlled Environment Gloves
SizeMedium
SterilityNon-sterile
Thickness (English)4 mil
Thickness (Metric)0.10 mm
Manufacturing/QMS Audit StandardsISO9001:2000 and ISO 14001:2004
ModelRVCR1502
Packaging TypeCleanroom Bagged
SeriesRVCR
Length (English)12 in.
Length (Metric)30.5 cm
Unit Size