Owl™ Transfer Membranes
Owl™ Transfer Membranes
Actual product may vary
Thermo Fisher Scientific
Owl™ Transfer Membranes
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPackaging Type
TM151-1Roll
TM151-4Sheets (15 x 15cm)
TM151-5Sheets (10 x 10cm)
Catalog number TM151-1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Packaging Type:
Roll
Perform Western blotting with hydrophobic Thermo Scientific™ Owl™ Transfer Membranes.
  • Thermo Scientific offers Immobilon™-P, PVDF transfer membrane manufactured by Millipore™.
  • This membrane provides the highest signal-to-noise ratio, which allows the use of mild blocking procedures.
  • Immobilon-P has high protein retention, is compatible with all standard visualization methods and can be used for protein sequencing.
  • Immobilon is available in minigel and large-format gel sizes.
Specifications
Length (Metric)26.5 cm
Width (Metric)3.75 m
Packaging TypeRoll
Description26.5cm x 3.75m; Roll
For Use With (Application)Western blotting
FormatRoll
MaterialImmobilon-P, PVDF
Dimensions (LxW)3.75 m x 26.5 cm
Product LineOwl
Unit Size