Phthalates in Polyvinyl Chloride Standard, 8 Components, SPEX CertiPrep™
Phthalates in Polyvinyl Chloride Standard, 8 Components, SPEX CertiPrep™
Phthalates in Polyvinyl Chloride Standard, 8 Components, SPEX CertiPrep™
Phthalates in Polyvinyl Chloride Standard, 8 Components, SPEX CertiPrep™
Phthalates in Polyvinyl Chloride Standard, 8 Components, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Have Questions?
Catalog NumberdimQuantity
SPPCRM-PVC001
Catalog number SPPCRM-PVC001
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Request bulk or custom format
Specifications
MethodsFor Consumer Product Safety Commission Method CPSC-CH-C1001-09.1
Unit Size
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Product intended for laboratory use only