Check Order Status
Replacement Tubes
Actual product may vary
Replacement Tubes
Have Questions?
Change view
Catalog Number
Wavelength
For Use With (Equipment)
SPLBLE-6254S
254 nm
E series, B series and miniMAX lamp
SPLBLE-6T365
365 nm
E series, B series and miniMAX lamp
2 Options
Catalog number
SPLBLE-6254S
Price (MYR)
2,003.00
Add to cart
Wavelength:
254 nm
For Use With (Equipment):
E series, B series and miniMAX lamp
Price (MYR)
2,003.00
Add to cart
Replacement Tubes
Specifications
Wattage
6 W
Wavelength
254 nm
For Use With (Equipment)
E series, B series and miniMAX lamp
Product Type
Replacement Tube
Unit Size