Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Actual product may vary
Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Thermo Fisher Scientific
Accessories for Solaris™ 2000 and 4000 Open Air Orbital Shakers
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionFor Use With (Equipment)Dimensions (L x W)
SK1824Solaris 18x24 Universal platformSolaris 200018 x 24 in.
SK1830Solaris 18x30 Universal platformSolaris 400018 x 30 in.
Catalog number SK1824
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Solaris 18x24 Universal platform
For Use With (Equipment):
Solaris 2000
Dimensions (L x W):
18 x 24 in.
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab.
Mix and match a variety of labware on a single platform with flexible Thermo Scientific™ Solaris™ Shaker Universal Platforms and Clamps. With exceptional innovation, reliability and accuracy, these accessories are the optimal choice for a busy lab. Select a platform and clamp configuration that best meets your application, whether you require a single vessel type and size.
Specifications
Product TypeUniversal Platform
Certifications/ComplianceCertificate of Analysis (COA), Certificate of Conformance (COC)
Dimensions (L x W)18 x 24 in.
Length (English) Platform18 in.
Length (Metric) Platform457.2 mm
Width (English) Platform24 in.
Width (Metric) Platform610 mm
DescriptionSolaris 18x24 Universal platform
For Use With (Equipment)Solaris 2000
IncludesPlatform
Warranty and Service Offering1 Year
Unit Size