MaxQ™ 4450 Benchtop Orbital Shakers
MaxQ™ 4450 Benchtop Orbital Shakers
Actual product may vary
Thermo Fisher Scientific
MaxQ™ 4450 Benchtop Orbital Shakers
Combine incubated and refrigerated temperatures with a small footprint by using these affordably-priced, orbital shakers, ideal for incubating a small number of vessels.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescriptionElectrical RequirementsSpeed RangeTemperature Range (Metric)
SHKE4450-1CESmall Incubated Benchtop Orbital Shaker240 V, 50/60 Hz15 to 500 rpm, ±1 rpmAmbient +5° to 80°C
SHKE4450CC-1CESmall Incubated Benchtop Orbital Shaker with cooling coil (required chiller)240 V, 50/60 Hz15 to 500 rpm, ±1 rpmAmbient -5° to 80°C
Catalog number SHKE4450-1CE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
Small Incubated Benchtop Orbital Shaker
Electrical Requirements:
240 V, 50/60 Hz
Speed Range:
15 to 500 rpm, ±1 rpm
Temperature Range (Metric):
Ambient +5° to 80°C
Combine incubated and refrigerated temperatures with a small footprint by using Thermo Scientific™ MaxQ™ 4450 Benchtop Orbital Shakers. These versatile shakers are ideal for incubating a small number of vessels and are used for cell culture, solubility studies, extraction procedures, protein expression and many other applications.
Key Features:
  • Clear, scratch-resistant lid allows unobstructed sample viewing without disturbing chamber temperature
  • Chamber holds four 1L flasks
  • Choose from two temperature ranges: incubated (5°C above ambient to 80°C) or with a cooling coil option that requires a chiller (5°C below ambient to 80°C)
  • Monitor and control chamber temperature range with ±0.1°C accuracy at 37°C
  • Triple eccentric drive handles heavy loads, uniform agitation and continuous 24-hour operation, even at high speeds
  • Overtemperature safety feature with independent thermostat provides additional backup by controlling heat if main temperature controller fails
  • Interlock stops platform when lid is opened
Analog Models:
  • Speed is controlled by a rotary dial, monitored accurately by a traditional tachometer
  • Easy-to-read LED digital temperature display
  • Speed: 40-400rpm Integrated tachometer monitors and displays speed in rpm to guarantee an accurate setting
  • Timed: 1 to 60 min. or continuous
Digital Models:
  • Three individual LED displays: speed, time and temperature
  • Variable speed control from 15 to 500rpm
  • Continuous and timed operation from 0.1 hour up to 999 hours or 0.1 to 999 min.
  • Shaker shutdown and audible/visual alarms signal temperature deviations or if unit operates ±10% of set speed, preventing shaker from “walking”
  • Soft-start feature eliminates sudden starts and stops, splashing of vessel contents or wetting of flask closure
  • Set point retention retains parameters during power failure and restarts unit automatically after power is restored
  • Solid-state DC brushless motor is maintenance-free
Operating Chiller:
  • Provides a refrigerated atmosphere
  • For plasmid purification and protein expression in bacteria for use with MaxQ 4450 with cooling coil option
  • Achieve temperature ranges from 20° to 30°C
Ordering Information:
For a complete system, order platforms and clamps (sold separately).
Requires:
Universal Platform (without clamps) or Dedicated Platform (with pre-installed clamps). All MaxQ 4450 shakers with cooling coil require a chiller sold separately.
Recommended for:
  • Cell culture
  • Solubility studies
  • Extraction procedures
  • Plasmid purification
  • General mixing
  • Bacterial suspensions
  • Staining and destaining
  • Washing procedures
  • Hybridizations
  • Protein expression in bacteria
Specifications
Orbit19 mm
Load Bearing Capacity (Metric)15.9 kg
Speed Range15 to 500 rpm, ±1 rpm
Electrical Requirements240 V, 50/60 Hz
Voltage220/240 V
Temperature Range (Metric)Ambient +5° to 80°C
Holds4 x 1 L Flasks
Product TypeShaker
Load Bearing Capacity (English)35 lb.
Certifications/ComplianceCE, cUL, UL
Temperature Range (English)Ambient + 9° F to 176° F
Temperature Accuracy± 0.1°C
Temperature Resolution±0.1°C
Frequency50/60 Hz
Dimensions (L x W x H)27.2 x 14.1 x 15.8 in. (69.1 x 35.8 x 40.1 cm)
Height (English)15.8 in.
Height (Metric)40.1 cm
Length (English)27.2 in.
Length (English) Platform11 in.
Length (Metric)69.1 cm
Length (Metric) Platform28 cm
Width (English)14.1 in.
Width (Metric)35.8 cm
Width (Metric) Platform33 cm
Shipping Weight (English)77 lb.
Shipping Weight (Metric)35 kg
Controller TypeDigital
DescriptionSmall Incubated Benchtop Orbital Shaker
ModelMaxQ 4450 Incubated Benchtop Shaker
Motor TypeDC brushless
No. of Positions4
Timer RangeContinuous or timed operation from 0.1 minute to 999 hours
Unit Size