eVol™ XCHANGE Syringes
eVol™ XCHANGE Syringes
Actual product may vary
Thermo Fisher Scientific
eVol™ XCHANGE Syringes
XCHANGE™ analytical syringes can be easily and quickly changed allowing them to be dedicated to individual liquids, and methods, to prevent possible cross-contamination of reagents.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Quantity
SGE29100351 mL1 Pk.
SGE2910329100 μL3/Pk.
Catalog number SGE2910035
Price (MYR)
835.00
Add to cart
Volume (Metric):
1 mL
Quantity:
1 Pk.
Price (MYR)
835.00
Add to cart
  • Liquid handling user independent device
  • Reduction in need for re-analysis
  • Syringes easily and quickly changed
  • Replacement parts available
Specifications
Needle Gauge23 G
For Use With (Application)Liquid handling procedure
Length (English)1.9 in.
Length (Metric)50 mm
Product TypeeVol™ Syringe
Quantity1 Pk.
Volume (Metric)1 mL
Length (Metric) Needle50 mm
Needle Point StyleBevel
Unit Size