Ig Goat F(ab')2 anti-Mouse, Human ads, AF 488, Polyclonal, Southern Biotech™
Ig Goat F(ab')2 anti-Mouse, Human ads, AF 488, Polyclonal, Southern Biotech™
Actual product may vary
Ig Goat F(ab')2 anti-Mouse, Human ads, AF 488, Polyclonal, Southern Biotech™
Goat F(ab')2 Polyclonal Secondary Antibody
Have Questions?
Catalog NumberQuantity
SGE1012300.5 mg
Catalog number SGE101230
Price (MYR)
235.00
EA
Add to cart
Quantity:
0.5 mg
Price (MYR)
235.00
EA
Add to cart
SouthernBiotech is a leading provider of polyclonal secondary antibodies offering high sensitivity and low background for use in a variety of applications.
Specifications
AntigenIg
ApplicationsFlow Cytometry, ELISA, Immunocytochemistry, Western Blot
ClassificationPolyclonal
Concentration0.5mg/mL
ConjugateAlexa Fluor 488
FormulationSupplied in PBS/NaN3
Host SpeciesGoat
ImmunogenMouse IgG, IgM, and IgA
Purification MethodAffinity Purified
Quantity0.5 mg
Regulatory StatusRUO
Primary or SecondarySecondary
Target SpeciesMouse
IsotypeIgG
Unit SizeEA
Contents & Storage
Store at 2-8°C; Protect from light