Check Order Status
GC Deactivated Inlet Liner Split/Splitless with Single Taper Quartz Wool For Agilent 7890
Thermo Fisher Scientific
GC Deactivated Inlet Liner Split/Splitless with Single Taper Quartz Wool For Agilent 7890
The unique deactivation process of SGE inlet liners provides excellent analysis for a wide range of compounds
Have Questions?
Catalog number
SGE09201901
Price (MYR)
235.00
Add to cart
Price (MYR)
235.00
Add to cart
Each batch of liners tested for activity
Manufactured from borosilicate glass
Color coded by geometry for easy identification
For use with Agilent instruments
Specifications
Diameter (Metric) Inner
4 mm
Length (Metric)
78.5 mm
Type
Split/Splitless with Single Taper (Quartz Wool) Inlet Liner
Diameter (Metric) Outer
6.3 mm
Unit Size