SilTite™ GC Capillary Column Connectors: μ Unions
SilTite™ GC Capillary Column Connectors: μ Unions
Actual product may vary
Thermo Fisher Scientific
SilTite™ GC Capillary Column Connectors: μ Unions
SilTite μ-Union delivers an easy, leak-free installation for capillary columns without the use of any tools
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConnections
SGE0735620.1-0.25 mm and 0.53 mm
SGE0735600.1-0.25 mm and 0.1-0.25 mm
Catalog number SGE073562
Price (MYR)
1,097.00
EA
Add to cart
Connections:
0.1-0.25 mm and 0.53 mm
Price (MYR)
1,097.00
EA
Add to cart
  • Low thermal mass
  • Zero Dead Volume
  • Inert
  • No-need to retighten after installation
  • Connect Pre-column or GC x GC
Specifications
TypeConnector
Connections0.1-0.25 mm and 0.53 mm
Includes2 unions, 5 double taper ferrules and tools
For Use With (Equipment)GC Capillary Column
Unit SizeEA