Gastight Syringes: Fixed Luer-Lok Models
Gastight Syringes: Fixed Luer-Lok Models
Actual product may vary
Gastight Syringes: Fixed Luer-Lok Models
Gastight Syringes: Fixed Luer-Lok Models
Thermo Fisher Scientific
Gastight Syringes: Fixed Luer-Lok Models
Suitable for liquid and gas samples
Have Questions?
Change viewbuttonViewtableView
Catalog NumberVolume (Metric)Diameter (English) OuterModel
SGE0084252.5 mL0.42 in.2.5MDF-LL-GT
SGE0080251 mL0.31 in.1MDF-LL-GT
Catalog number SGE008425
Price (MYR)
527.00
Add to cart
Volume (Metric):
2.5 mL
Diameter (English) Outer:
0.42 in.
Model:
2.5MDF-LL-GT
Price (MYR)
527.00
Add to cart
  • 10μL to 100mL capacities
  • Constructed from borosilicate glass, with PTFE plunger tips and Kel-F™ Luer cones (where applicable)
  • Needles align with zero mark on barrel to reduce dead volume, improve accuracy and precision to <±1%
  • Replaceable plungers on all models
  • Luer-tip and Luer-Lok™ syringes come without needles; order separately
Specifications
Accuracy±1%
MaterialBorosilicate Glass (Barrel), PTFE (Tip)
Diameter (English) Outer0.42 in.
Diameter (Metric) Outer10.8 mm
For Use With (Application)Suitable for gases or liquids for maximum inertness
IncludesPlunger protection, inert Kel-F male luer cone
Product TypeGastight Syringe
Max. Temperature (English)158°F
Max. Temperature (Metric)70°C
Model2.5MDF-LL-GT
Needle Point NumberNo Needle Included
QuantityEa.
Volume (Metric)2.5 mL
Connection TypeLuer Lock, Fixed
Unit Size