Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
Actual product may vary
Autosampler Syringes with Removable Needle for CTC Analytics CombiPAL and GC PAL
For reduced non specific sample interaction and increased analyte recovery
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNeedle GaugeDiameter (English) Outer NeedleLength (English) NeedleModel
SGE00386626 G0.01 in.2.2 in.25F-RTC/RSH-GT-5.7/0.47C
SGE00286526 G0.01 in.2.2 in.10F-RTC/RSH-5.7/0.47C
Catalog number SGE003866
Price (MYR)
920.00
EA
Add to cart
Needle Gauge:
26 G
Diameter (English) Outer Needle:
0.01 in.
Length (English) Needle:
2.2 in.
Model:
25F-RTC/RSH-GT-5.7/0.47C
Price (MYR)
920.00
EA
Add to cart
  • Borosilicate glass barrel for strength and inertness.
  • Stainless steel needles for inertness, strength and corrosion resistance.
  • Backing behind the scale aids quick and accurate reading of the syringe scale.
  • Metal back flange provides stability and resists fracture.
  • Metal needle cap on syringe barrel protects and improves syringe lifetime.
  • Syringes are individually tested for quality ensuring reliability and conformance.
  • Unique packaging design eliminates possible contamination in the syringe.
Specifications
Needle Gauge26 G
Diameter (English) Outer Needle0.01 in.
Diameter (Metric) Outer Needle0.47 mm
For Use With (Application)Gas chromatography
Length (English) Needle2.2 in.
Product TypeAutosampler Syringe
Model25F-RTC/RSH-GT-5.7/0.47C
QuantityEa.
Volume (Metric)25 μL
Needle Point StyleCone
For Use With (Equipment)Thermo RSH Autosampler
Connection TypeFixed
Unit SizeEA