Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Actual product may vary
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Thermo Fisher Scientific
Savant™ DNA SpeedVac™ Concentrators Miscellaneous Accessories
Improve the functionality of Savant SpeedVac Concentrators with these Savant SpeedVac Concentrators miscellaneous accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
SCC5CryoCool Heat Transfer Fluid, 5L
SCT120Chemical Trap (order cartridges separately)
GCF400Glass condensation flask
Catalog number SCC5
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Description:
CryoCool Heat Transfer Fluid, 5L
Improve the funcionality of Thermo Scientific™ Savant™ SpeedVac™ Concentrators with Savant™ SpeedVac™ Concentrators Accessories.
Specifications
DescriptionCryoCool Heat Transfer Fluid, 5L
TypeHeat Transfer Fluid
For Use With (Equipment)Refrigerated Trap Models RVT100, RVT400, RVT405DDA, RVT5105 and Universal Vacuum Source (UVS) Models UVS400, USV400A, UVS800DDA
ModelSCC5
Volume (Metric)5 L
Unit Size