Isotemp™ Stirrer
Isotemp™ Stirrer
Actual product may vary
Thermo Fisher Scientific
Isotemp™ Stirrer
Experience precise stirring control and exceptional safety performance for your routine protocols. Available in three sizes, which provide flexibility from microscale chemistries to production operations.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTop Plate MaterialElectrical RequirementsTop Plate Surface Area (English)Voltage
S88850205Ceramic230V 50/60Hz10.25 x 10.25 in.230 V
S88854205Ceramic230V 50/60Hz4.25 x 4.25 in.230 V
S88857205Ceramic230V 50/60Hz7.25 x 7.25 in.230 V
Catalog number S88850205
Price (MYR)
3,643.00
Add to cart
Top Plate Material:
Ceramic
Electrical Requirements:
230V 50/60Hz
Top Plate Surface Area (English):
10.25 x 10.25 in.
Voltage:
230 V
Price (MYR)
3,643.00
Add to cart
Experience precise stirring control and exceptional safety performance for your routine protocols. Available in three sizes, which provide flexibility from microscale chemistries to production operations.
Advanced technology
  • Slow-speed stirring, consistent speed control, and strong magnetic coupling
Flexible, robust design
  • Top plate choices in ceramic or aluminum
  • Raised display design protects electronics from spills
  • Optional splash guard protection shield
  • Variable sizes for different sample volumes
  • Includes stir bar
Specifications
Speed50 to 1500 rpm
Top Plate MaterialCeramic
Voltage230 V
Dimensions (L x W x H)16.2 x 11.3 x 4 in. (41.1 x 28.7 x 10.2 cm)
Top Plate Surface Area (English)10.25 x 10.25 in.
Top Plate Surface Area (Metric)26.0 x 26.0 cm
Electrical Requirements230V 50/60Hz
Frequency50/60 Hz
Plug TypeUK/EU/CN
DescriptionStirrer
Unit Size