Search Thermo Fisher Scientific
Catalog Number | Volume (Metric) Buffer | Includes |
---|---|---|
P9DS-1 | 550 mL | Complete System plus Caster |
P10DS-1 | 1250 mL | Complete System plus Caster |
P8DS-1 | 300 mL | Complete System plus Caster |
P8DS-2 | 300 mL | Complete System |
P9DS-2 | 550 mL | Complete System |
Quickly and easily cast and run one or two gels in the same device using Thermo Scientific™ Owl™ Dual-Gel Vertical Electrophoresis Systems—comprised of the P8DS, P9DS and P10DS Dual Gel Systems.
Simple, Six-Step Use:
Features:
Recommended for: