Starter Water Analysis Meter Replacement Electrodes
Starter Water Analysis Meter Replacement Electrodes
Actual product may vary
Starter Water Analysis Meter Replacement Electrodes
Replacement electrodes for Starter portable and benchtop meters.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelConnector TypesLength (Metric)Type
OHA83033965Model ST310BNC/Cinch120 mm3-in-1 Plastic Refillable pH Electrode
OHA83033966Model ST210BNC120 mm2-in-1 Plastic Refillable pH Electrode
OHA83033968Model ST230BNC/Cinch110 mm2-in 1 Glass Muddy Sample pH Electrode
OHA83033967Model ST320BNC120 mm3-in-1 Plastic Gel pH Electrode
Catalog number OHA83033965
Price (MYR)
729.00
Add to cart
Model:
Model ST310
Connector Types:
BNC/Cinch
Length (Metric):
120 mm
Type:
3-in-1 Plastic Refillable pH Electrode
Price (MYR)
729.00
Add to cart
Compatible Meters
  • Starter benchtop: 2100, 3100 and 3100C
  • Starter portable: 300, 300C and 300D
Specifications
For Use With (Equipment)Starter 300 pH meter
Sensor TypespH Electrode
Temperature (Metric) Range0 to 80°C
Cell Constant0.1 μS/cm
Type3-in-1 Plastic Refillable pH Electrode
Connector TypesBNC/Cinch
Diameter (English) Body0.47 in.
Diameter (Metric) Body12 mm
Length (English) Cable39.3 in.
Length (Metric)120 mm
Length (Metric) Cable1 m
MaterialPlastic
Measurement RangespH 0 to 14
Filling Solution Type3 mol/L KCl gel
Internal ReferenceAg/AgCl
JunctionCeramic Pin
ModelModel ST310
Unit Size