Stainless Steel Calibration Weights - OIML F1, NVLAP
Stainless Steel Calibration Weights - OIML F1, NVLAP
Actual product may vary
Stainless Steel Calibration Weights - OIML F1, NVLAP
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F1 Tolerances
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMass
OHA8078033650 mg
OHA80780337100 mg
Catalog number OHA80780336
Price (MYR)
953.00
Add to cart
Mass:
50 mg
Price (MYR)
953.00
Add to cart
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifications
Certifications/ComplianceNVLAP Accredited Certificate
MaterialStainless Steel (304)
Weight (Metric)0.030 kg
Mass50 mg
Tolerance0.040 mg
IncludesProtective Case
Unit Size