Conductivity Standards
Conductivity Standards
Actual product may vary
Conductivity Standards
Measure the ability of an electrolyte solution to conduct electricity.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberConductivity
OHA3010044284 μS/cm
OHA301004431413 μS/cm
Catalog number OHA30100442
Price (MYR)
169.00
Add to cart
Conductivity:
84 μS/cm
Price (MYR)
169.00
Add to cart
  • 250 mL Solution
  • Available in values of 84 μs/cm, 1413 μs/cm and 12.88 μs/cm
Extremes of temperature and direct sunlight should be avoided.
Specifications
Boiling Point∼100°C
Capacity (English)8.45 oz.
Capacity (Metric)250 mL
ColorClear, Colorless
Conductivity84 μS/cm
For Use With (Equipment)Conductivity Sensor
TypeConductivity Standards
pH Range∼6
Water SolubleCompletely Miscible
Unit Size